| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000484918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000484918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000484918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000486931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000486931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000486931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
ENST00000486931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11001104.hg.1 |
ENST00000381514.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11001104.hg.1 |
ENST00000381519.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11001104.hg.1 |
ENST00000397294.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11001104.hg.1 |
ENST00000397297.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11001104.hg.1 |
ENST00000397298.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11001104.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11001104.hg.1 |
TC11000055.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC11_KI270721v1_random00006432.hg.1 |
ENST00000612848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000484918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000484918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000484918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000486931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000486931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000486931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
ENST00000486931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006547.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000381514.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000381519.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000397294.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000397297.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000397298.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000429295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000462288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
ENST00000466346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
NM_021134.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100145758.hg.1 |
TC1100006547.hg.1 |
Clariom D TC |