| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017331.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017331.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017331.hg.1 |
TC0X001279.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010515.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171696.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171696.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171696.hg.1 |
ENST00000324068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171696.hg.1 |
NM_014289.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171696.hg.1 |
TC0X00010515.hg.1 |
Clariom D TC |