Assay Details
| Gene Symbol: OSGIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126111.2 | NP_001119583.1 | 2 - 3 | 460 | 116 | |
| NM_004337.2 | NP_004328.1 | 2 - 3 | 414 | 116 | ||
| XM_011517287.2 | XP_011515589.1 | 2 - 3 | 322 | 116 | ||
| GenBank mRNA | AF061326.1 | - | 2 - 3 | 413 | 116 | |
| BC031054.1 | - | 2 - 3 | 458 | 116 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000546.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007455.hg.1 | ENST00000451899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007455.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007455.hg.1 | TC08000546.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007458.hg.1 | ENST00000297438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007458.hg.1 | ENST00000451899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007458.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007458.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007458.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007458.hg.1 | TC08000546.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007459.hg.1 | ENST00000297438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007459.hg.1 | ENST00000451899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007459.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007459.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007459.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007459.hg.1 | TC08000546.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007456.hg.1 | ENST00000451899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007456.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007456.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007456.hg.1 | TC08000546.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800156161.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156161.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156161.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156161.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156161.hg.1 | TC0800008183.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800156162.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156162.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156162.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156162.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156162.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156162.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156162.hg.1 | TC0800008183.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800156163.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156163.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156163.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156163.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156163.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156163.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156163.hg.1 | TC0800008183.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156165.hg.1 | TC0800008183.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008183.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | ENST00000297438.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | ENST00000451899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | ENST00000520659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | NM_001126111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | NM_004337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156166.hg.1 | TC0800008183.hg.1 | Clariom D TC |

OSGIN2
Genomic Map