Assay Details
| Gene Symbol: FOXRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102371.1 | NP_001095841.1 | 5 - 6 | 1307 | 150 | |
| NM_024955.5 | NP_079231.4 | 5 - 6 | 1346 | 150 | ||
| XM_017028941.1 | XP_016884430.1 | 4 - 5 | 979 | 150 | ||
| XM_017028942.1 | XP_016884431.1 | 4 - 5 | 1076 | 150 | ||
| GenBank mRNA | AK025714.1 | - | 5 - 6 | 1246 | 150 | |
| AK026975.1 | - | 2 - 3 | 427 | 150 | ||
| AK225347.1 | - | 5 - 6 | 1246 | 150 | ||
| AK315555.1 | - | 5 - 6 | 1249 | 150 | ||
| BC015726.1 | - | 5 - 6 | 1230 | 4866761 | 150 | |
| BC027716.2 | - | 5 - 6 | 1325 | 4127473 | 150 | |
| BC040351.1 | - | 5 - 6 | 1269 | 3873448 | 150 | |
| CR456444.1 | - | 4 - 5 | 1321 | 150 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000710.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22012499.hg.1 | ENST00000216187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012499.hg.1 | ENST00000366463.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012499.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012499.hg.1 | ENST00000397224.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012499.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012499.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012499.hg.1 | TC22000710.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012501.hg.1 | ENST00000216187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012501.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012501.hg.1 | ENST00000397224.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012501.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012501.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012501.hg.1 | TC22000710.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | TC2200008610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162028.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | TC2200008610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162030.hg.1 | NM_024955.5 | Refseq |

FOXRED2
Genomic Map