Assay Details
| Gene Symbol: CYB561A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161452.1 | NP_001154924.1 | 2 - 3 | 570 | 64 | |
| NM_001161454.1 | NP_001154926.1 | 5 - 6 | 1225 | 64 | ||
| NM_001300763.1 | NP_001287692.1 | 4 - 5 | 1056 | 64 | ||
| NM_153611.4 | NP_705839.3 | 4 - 5 | 1056 | 64 | ||
| XM_011544821.1 | XP_011543123.1 | 5 - 6 | 1241 | 64 | ||
| XM_011544823.1 | XP_011543125.1 | 3 - 4 | 976 | 64 | ||
| XM_017017345.1 | XP_016872834.1 | 6 - 7 | 3158 | 64 | ||
| XM_017017346.1 | XP_016872835.1 | 5 - 6 | 2989 | 64 | ||
| GenBank mRNA | AK056751.1 | - | 4 - 5 | 1047 | 64 | |
| AK075559.1 | - | 4 - 5 | 657 | 64 | ||
| AK225754.1 | - | 2 - 3 | 433 | 64 | ||
| AK297084.1 | - | 3 - 4 | 486 | 64 | ||
| BC014045.2 | - | 4 - 5 | 866 | 3503330 | 64 | |
| BC047710.1 | - | 4 - 5 | 575 | 6018124 | 64 | |
| DC389589.1 | - | 3 - 4 | 474 | 64 | ||
More Information
Set Membership: |
|

CYB561A3
Genomic Map