| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008815.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008815.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008815.hg.1 |
TC20000610.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158805.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158805.hg.1 |
NM_017545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158805.hg.1 |
TC2000008344.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158805.hg.1 |
ENST00000378789.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158805.hg.1 |
NM_017545.2 |
Refseq |