Assay Details
| Gene Symbol: PON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000446.5 | NP_000437.3 | 5 - 6 | 591 | 126 | |
| GenBank mRNA | AK314027.1 | - | 5 - 6 | 517 | 126 | |
| BC074719.2 | - | 5 - 6 | 523 | 30915169 | 126 | |
| CB122285.1 | - | 5 - 6 | 551 | 126 | ||
| D84371.1 | - | 5 - 6 | 548 | 126 | ||
| M63012.1 | - | 5 - 6 | 503 | 126 | ||
| M63013.1 | - | 5 - 6 | 442 | 126 | ||
| M63014.1 | - | 4 - 5 | 408 | 126 | ||
| S64615.1 | - | 5 - 6 | 503 | 126 | ||
| U53784.1 | - | 5 - 6 | 548 | 126 | ||
| Z70723.1 | - | 5 - 6 | 548 | 126 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003363.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cholesterol metabolic process
response to external stimulus
response to toxic substance
positive regulation of cholesterol efflux
dephosphorylation
aromatic compound catabolic process
response to nutrient levels
positive regulation of transporter activity
negative regulation of plasma lipoprotein particle oxidation
carboxylic acid catabolic process
organophosphate catabolic process
phosphatidylcholine metabolic process
positive regulation of binding
response to fatty acid
response to fluoride
aryldialkylphosphatase activity
arylesterase activity
calcium ion binding
phospholipid binding
protein homodimerization activity
acyl-L-homoserine-lactone lactonohydrolase activity
cholesterol metabolic process
response to external stimulus
response to toxic substance
positive regulation of cholesterol efflux
dephosphorylation
aromatic compound catabolic process
response to nutrient levels
positive regulation of transporter activity
negative regulation of plasma lipoprotein particle oxidation
carboxylic acid catabolic process
organophosphate catabolic process
phosphatidylcholine metabolic process
positive regulation of binding
response to fatty acid
response to fluoride
aryldialkylphosphatase activity
arylesterase activity
calcium ion binding
phospholipid binding
protein homodimerization activity
acyl-L-homoserine-lactone lactonohydrolase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07024524.hg.1 | ENST00000222381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024524.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024524.hg.1 | TC07003363.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024522.hg.1 | ENST00000222381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024522.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024522.hg.1 | TC07003363.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024523.hg.1 | ENST00000222381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024523.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024523.hg.1 | TC07003363.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700184538.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184538.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184538.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700184538.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184538.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184538.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700184538.hg.1 | TC0700011831.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000462594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011831.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700184539.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184539.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184539.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700184539.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184539.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184539.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700184539.hg.1 | TC0700011831.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700184540.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184540.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184540.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184540.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700184540.hg.1 | ENST00000222381.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184540.hg.1 | ENST00000433729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184540.hg.1 | ENST00000470502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700184540.hg.1 | NM_000446.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700184540.hg.1 | TC0700011831.hg.1 | Clariom D TC |

PON1
Genomic Map