Assay Details
| Gene Symbol: TSNAXIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288991.1 | NP_001275920.1 | 3 - 4 | 550 | 73 | |
| NM_001288993.1 | NP_001275922.1 | 3 - 4 | 550 | 73 | ||
| XM_011523236.2 | XP_011521538.1 | 3 - 4 | 320 | 73 | ||
| XM_011523241.1 | XP_011521543.1 | 3 - 4 | 510 | 73 | ||
| XM_017023445.1 | XP_016878934.1 | 3 - 4 | 320 | 73 | ||
| XM_017023447.1 | XP_016878936.1 | 3 - 4 | 320 | 73 | ||
| XM_017023449.1 | XP_016878938.1 | 3 - 4 | 320 | 73 | ||
| XM_017023453.1 | XP_016878942.1 | 3 - 4 | 510 | 73 | ||
| GenBank mRNA | AF132730.1 | - | 3 - 4 | 354 | 73 | |
| AK303841.1 | - | 3 - 4 | 340 | 73 | ||
| DA200633.1 | - | 3 - 4 | 510 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF132730.1, NM_001288993.1, XM_011523241.1, XM_017023453.1 transcripts.
Set Membership: |
|

TSNAXIP1
Genomic Map