Assay Details
| Gene Symbol: DCAF16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006713966.3 | XP_006714029.1 | 1 - 2 | 317 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006713966.3 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |

DCAF16
Genomic Map