Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | granulysin | GNLY |
D2S69E, LAG-2, LAG2, NKG5, TLA519 |
-12.6kb | Sense | Chr. 2 - 85694291 - 85698854 on NCBI Build GRCh38 |
| Downstream | atonal bHLH transcription factor 8 | ATOH8 |
HATH6, bHLHa21 |
41.6kb | Sense | Chr. 2 - 85753786 - 85791383 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map