Assay Details
| Gene Symbol: ZNF785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152458.6 | NP_689671.2 | 1 - 2 | 362 | 63 | |
| XM_017022967.1 | XP_016878456.1 | 1 - 2 | 629 | 63 | ||
| GenBank mRNA | AK056692.1 | - | 1 - 2 | 363 | 63 | |
| AK292465.1 | - | 1 - 2 | 246 | 63 | ||
| BC040642.2 | - | 1 - 2 | 280 | 4811661 | 63 | |
| CN401082.1 | - | 1 - 2 | 362 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001038.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16014835.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014835.hg.1 | ENST00000395222.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014835.hg.1 | ENST00000470110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014835.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014835.hg.1 | TC16001038.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014837.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014837.hg.1 | ENST00000470110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014837.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014837.hg.1 | TC16001038.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014833.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014833.hg.1 | ENST00000395222.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014833.hg.1 | ENST00000470110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014833.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014833.hg.1 | TC16001038.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014836.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014836.hg.1 | ENST00000395222.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014836.hg.1 | ENST00000470110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014836.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014836.hg.1 | TC16001038.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600177152.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177152.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177152.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177152.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177152.hg.1 | TC1600009978.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600177152.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177152.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177152.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177152.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177154.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177154.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177154.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177154.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177154.hg.1 | TC1600009978.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600177154.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177154.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177154.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177154.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177157.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177157.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177157.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177157.hg.1 | TC1600009978.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600177157.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177157.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177157.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177155.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177155.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177155.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177155.hg.1 | TC1600009978.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600177155.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177155.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177155.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177156.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177156.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177156.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177156.hg.1 | TC1600009978.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600177156.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177156.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177156.hg.1 | NM_152458.6 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 111511 | - | - |

ZNF785
Genomic Map