Assay Details
| Gene Symbol: ZNF785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011545753.2 | XP_011544055.1 | 3 - 4 | 1729 | 119 | |
| XM_017022967.1 | XP_016878456.1 | 3 - 4 | 1690 | 119 | ||
| GenBank mRNA | AK056692.1 | - | 3 - 4 | 1379 | 119 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007477.hg.1 | ENST00000486926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007477.hg.1 | ENST00000486926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007477.hg.1 | ENST00000486926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007477.hg.1 | ENST00000492040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007477.hg.1 | ENST00000492040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007477.hg.1 | ENST00000492040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007477.hg.1 | ENST00000486926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007477.hg.1 | ENST00000486926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007477.hg.1 | ENST00000486926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007477.hg.1 | ENST00000492040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007477.hg.1 | ENST00000492040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007477.hg.1 | ENST00000492040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |

ZNF785
Genomic Map