Assay Details
| Gene Symbol: N4BP2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286461.1 | NP_001273390.1 | 5 - 6 | 624 | 131 | |
| GenBank mRNA | BF448179.1 | - | 5 - 6 | 461 | 131 | |
More Information
Set Membership: |
|

N4BP2L1
Genomic Map