Assay Details
| Gene Symbol: RTF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015138.4 | NP_055953.3 | 7 - 8 | 1042 | 81 | |
| GenBank mRNA | AK292302.1 | - | 7 - 8 | 1014 | 81 | |
| BC015052.2 | - | 7 - 8 | 992 | 3909737 | 81 | |
| D87440.2 | - | 7 - 8 | 1008 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000308.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate commitment
blastocyst growth
transcription elongation from RNA polymerase II promoter
Wnt signaling pathway
stem cell population maintenance
positive regulation of transcription elongation from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
histone H3-K4 trimethylation
single-stranded DNA binding
protein binding
poly(A) RNA binding
negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate commitment
blastocyst growth
transcription elongation from RNA polymerase II promoter
Wnt signaling pathway
stem cell population maintenance
positive regulation of transcription elongation from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
histone H3-K4 trimethylation
single-stranded DNA binding
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15002240.hg.1 | ENST00000389629.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002240.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002240.hg.1 | TC15000308.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000558298.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000558298.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000558298.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000561076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000561076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000561076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | ENST00000561076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000462276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000473214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000558117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000558298.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000558298.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000558298.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000560447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000561076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000561076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000561076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | ENST00000561076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007004.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149003.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149003.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149003.hg.1 | ENST00000389629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149003.hg.1 | NM_015138.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149003.hg.1 | TC1500007004.hg.1 | Clariom D TC |

RTF1
Genomic Map