Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004251.4 | NP_004242.1 | 2 - 3 | 256 | 118 | |
| XM_017029964.1 | XP_016885453.1 | 2 - 3 | 658 | 118 | ||
| GenBank mRNA | BC017265.2 | - | 2 - 3 | 155 | 4139714 | 118 |
| DA141710.1 | - | 2 - 3 | 281 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC017265.2,NM_004251.4,XM_017029964.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

RAB9A
Genomic Map