Assay Details
| Gene Symbol: NUP62CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017681.2 | NP_060151.2 | 6 - 7 | 661 | 143 | |
| NR_033676.1 | 4 - 5 | 466 | 143 | |||
| GenBank mRNA | AK000137.1 | - | 6 - 7 | 535 | 143 | |
| BC016327.1 | - | 6 - 7 | 658 | 4102213 | 143 | |
| BC017799.1 | - | 4 - 5 | 459 | 143 | ||
| KU178655.1 | - | 4 - 5 | 402 | 143 | ||
| KU178656.1 | - | 4 - 5 | 402 | 143 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001260.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017027.hg.1 | ENST00000372461.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017027.hg.1 | ENST00000372465.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017027.hg.1 | ENST00000372466.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017027.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017027.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017027.hg.1 | TC0X001260.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171293.hg.1 | TC0X00010465.hg.1 | Clariom D TC |

NUP62CL
Genomic Map