Assay Details
| Gene Symbol: EIF2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004094.4 | NP_004085.1 | 5 - 6 | 719 | 120 | |
| GenBank mRNA | AK312442.1 | - | 5 - 6 | 708 | 120 | |
| BC002513.2 | - | 5 - 6 | 668 | 3139624 | 120 | |
| BX161521.1 | - | 5 - 6 | 704 | 120 | ||
| J02645.1 | - | 5 - 6 | 605 | 120 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000406.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
translational initiation
aging
negative regulation of translational initiation in response to stress
cellular response to amino acid starvation
cellular response to heat
cellular response to UV
response to endoplasmic reticulum stress
PERK-mediated unfolded protein response
protein autophosphorylation
transmembrane transport
mitophagy in response to mitochondrial depolarization
xenophagy
positive regulation of neuron death
response to manganese-induced endoplasmic reticulum stress
positive regulation of type B pancreatic cell apoptotic process
translation initiation factor activity
protein binding
ribosome binding
poly(A) RNA binding
positive regulation of defense response to virus by host
translational initiation
aging
negative regulation of translational initiation in response to stress
cellular response to amino acid starvation
cellular response to heat
cellular response to UV
response to endoplasmic reticulum stress
PERK-mediated unfolded protein response
protein autophosphorylation
transmembrane transport
mitophagy in response to mitochondrial depolarization
xenophagy
positive regulation of neuron death
response to manganese-induced endoplasmic reticulum stress
positive regulation of type B pancreatic cell apoptotic process
translation initiation factor activity
protein binding
ribosome binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000554332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000554332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000554332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000554332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004763.hg.1 | ENST00000256383.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004763.hg.1 | ENST00000466499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004763.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004763.hg.1 | TC14000406.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000554332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000554332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000554332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000554332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000556724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007494.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | ENST00000256383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | ENST00000466499.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | ENST00000555876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | ENST00000557310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | NM_004094.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152514.hg.1 | TC1400007494.hg.1 | Clariom D TC |

EIF2S1
Genomic Map