| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026506.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026506.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026506.hg.1 |
TC06002112.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026505.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026505.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026505.hg.1 |
TC06002112.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600191044.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600191044.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600191044.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600191044.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600191044.hg.1 |
TC0600013186.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600191042.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600191042.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600191042.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600191042.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600191042.hg.1 |
TC0600013186.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600191043.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600191043.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600191043.hg.1 |
ENST00000367976.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600191043.hg.1 |
NM_001901.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600191043.hg.1 |
TC0600013186.hg.1 |
Clariom D TC |