Assay Details
| Gene Symbol: ZNF326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320185.1 | NP_001307114.1 | 11 - 12 | 79 | ||
| NM_181781.3 | NP_861446.2 | 9 - 10 | 79 | |||
| NM_182976.3 | NP_892021.1 | 11 - 12 | 79 | |||
| XM_005270779.4 | XP_005270836.1 | 10 - 11 | 79 | |||
| XM_011541290.2 | XP_011539592.1 | 11 - 12 | 79 | |||
| GenBank mRNA | AK297073.1 | - | 11 - 12 | 79 | ||
| AK303707.1 | - | 9 - 10 | 79 | |||
| BC014899.1 | - | 11 - 12 | 79 | |||
| BC070341.1 | - | 11 - 12 | 79 | |||
| BC090866.1 | - | 11 - 12 | 1556 | 6503526 | 79 | |
| BU521884.1 | - | 3 - 4 | 79 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000842.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01013392.hg.1 | ENST00000340281.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013392.hg.1 | ENST00000370447.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013392.hg.1 | ENST00000394590.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013392.hg.1 | ENST00000455342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013392.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013392.hg.1 | TC01000842.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013393.hg.1 | ENST00000340281.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013393.hg.1 | ENST00000370447.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013393.hg.1 | ENST00000394590.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013393.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013393.hg.1 | TC01000842.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013395.hg.1 | ENST00000340281.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013395.hg.1 | ENST00000370447.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013395.hg.1 | ENST00000394590.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013395.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013395.hg.1 | TC01000842.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100164174.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164174.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164174.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164174.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164174.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164174.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164174.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164174.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164174.hg.1 | TC0100008996.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164177.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164177.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164177.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164177.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164177.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164177.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164177.hg.1 | TC0100008996.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100164175.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164175.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164175.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164175.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164175.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164175.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164175.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164175.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164175.hg.1 | TC0100008996.hg.1 | Clariom D TC |

ZNF326
Genomic Map