Assay Details
| Gene Symbol: ZNF326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182976.3 | NP_892021.1 | 4 - 5 | 371 | 83 | |
| GenBank mRNA | AK297073.1 | - | 4 - 5 | 296 | 83 | |
| BC052645.1 | - | 4 - 5 | 345 | 83 | ||
| BC070341.1 | - | 4 - 5 | 315 | 83 | ||
| BC090866.1 | - | 4 - 5 | 365 | 6503526 | 83 | |
| DB111509.1 | - | 4 - 5 | 347 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000842.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01013384.hg.1 | ENST00000340281.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013384.hg.1 | ENST00000394590.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013384.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013384.hg.1 | TC01000842.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164166.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164166.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164166.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164166.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164166.hg.1 | TC0100008996.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |

ZNF326
Genomic Map