Assay Details
| Gene Symbol: ZNF326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320185.1 | NP_001307114.1 | 5 - 6 | 511 | 109 | |
| NM_182976.3 | NP_892021.1 | 5 - 6 | 778 | 109 | ||
| XM_011541290.2 | XP_011539592.1 | 5 - 6 | 484 | 109 | ||
| GenBank mRNA | AK297073.1 | - | 5 - 6 | 703 | 109 | |
| BC014899.1 | - | 5 - 6 | 469 | 109 | ||
| BC052645.1 | - | 5 - 6 | 752 | 109 | ||
| BC070341.1 | - | 5 - 6 | 722 | 109 | ||
| BC090866.1 | - | 5 - 6 | 772 | 6503526 | 109 | |
| CN299618.1 | - | 5 - 6 | 499 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000842.hg
Additional Information:
This assay is in the 5'UTR region of the XM_011541290.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013385.hg.1 | ENST00000340281.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013385.hg.1 | ENST00000370447.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013385.hg.1 | ENST00000394590.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013385.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013385.hg.1 | TC01000842.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013386.hg.1 | ENST00000340281.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013386.hg.1 | ENST00000370447.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013386.hg.1 | ENST00000394590.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013386.hg.1 | ENST00000455342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013386.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013386.hg.1 | TC01000842.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100164168.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164168.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164168.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164168.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164168.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164168.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164168.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164168.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164168.hg.1 | TC0100008996.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100164167.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164167.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164167.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164167.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164167.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164167.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164167.hg.1 | TC0100008996.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |

ZNF326
Genomic Map