| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013387.hg.1 |
ENST00000340281.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013387.hg.1 |
ENST00000370447.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013387.hg.1 |
ENST00000394590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013387.hg.1 |
ENST00000455342.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013387.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013387.hg.1 |
TC01000842.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000361911.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182975.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008996.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100164169.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164169.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164169.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164169.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100164169.hg.1 |
ENST00000340281.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164169.hg.1 |
ENST00000370447.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164169.hg.1 |
ENST00000394583.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164169.hg.1 |
NM_182976.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100164169.hg.1 |
TC0100008996.hg.1 |
Clariom D TC |