Assay Details
| Gene Symbol: ZNF326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320185.1 | NP_001307114.1 | 7 - 8 | 813 | 138 | |
| NM_181781.3 | NP_861446.2 | 5 - 6 | 616 | 138 | ||
| NM_182976.3 | NP_892021.1 | 7 - 8 | 1080 | 138 | ||
| XM_005270779.4 | XP_005270836.1 | 6 - 7 | 654 | 138 | ||
| XM_011541290.2 | XP_011539592.1 | 7 - 8 | 786 | 138 | ||
| GenBank mRNA | AK297073.1 | - | 7 - 8 | 1005 | 138 | |
| AK303707.1 | - | 5 - 6 | 543 | 138 | ||
| BC014899.1 | - | 7 - 8 | 771 | 138 | ||
| BC070341.1 | - | 7 - 8 | 1024 | 138 | ||
| BC090866.1 | - | 7 - 8 | 1074 | 6503526 | 138 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000842.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013388.hg.1 | ENST00000340281.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013388.hg.1 | ENST00000370447.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013388.hg.1 | ENST00000394590.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013388.hg.1 | ENST00000455342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013388.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013388.hg.1 | TC01000842.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164170.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164170.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164170.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164170.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164170.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164170.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164170.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164170.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164170.hg.1 | TC0100008996.hg.1 | Clariom D TC |

ZNF326
Genomic Map