Assay Details
| Gene Symbol: ZNF326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320185.1 | NP_001307114.1 | 8 - 9 | 968 | 139 | |
| NM_181781.3 | NP_861446.2 | 6 - 7 | 771 | 139 | ||
| NM_182976.3 | NP_892021.1 | 8 - 9 | 1235 | 139 | ||
| XM_005270779.4 | XP_005270836.1 | 7 - 8 | 809 | 139 | ||
| XM_011541290.2 | XP_011539592.1 | 8 - 9 | 941 | 139 | ||
| GenBank mRNA | AK297073.1 | - | 8 - 9 | 1160 | 139 | |
| AK303707.1 | - | 6 - 7 | 698 | 139 | ||
| BC014899.1 | - | 8 - 9 | 926 | 139 | ||
| BC070341.1 | - | 8 - 9 | 1179 | 139 | ||
| BC090866.1 | - | 8 - 9 | 1229 | 6503526 | 139 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000842.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013389.hg.1 | ENST00000340281.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013389.hg.1 | ENST00000370447.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013389.hg.1 | ENST00000394590.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013389.hg.1 | ENST00000455342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013389.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013389.hg.1 | TC01000842.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000361911.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182975.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008996.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164171.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164171.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164171.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164171.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164171.hg.1 | ENST00000340281.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164171.hg.1 | ENST00000370447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164171.hg.1 | ENST00000394583.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164171.hg.1 | NM_182976.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164171.hg.1 | TC0100008996.hg.1 | Clariom D TC |

ZNF326
Genomic Map