| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000473432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000473432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000473432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000473432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025158.hg.1 |
ENST00000305997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025158.hg.1 |
ENST00000486484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025158.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025158.hg.1 |
TC02001556.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200225676.hg.1 |
TC0200016626.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000460786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000462696.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000471753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000473432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000473432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000473432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000473432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000474341.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000477073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486315.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000486484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
ENST00000494760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016626.hg.1 |
NM_138799.3 |
Refseq |