Assay Details
| Gene Symbol: CHAF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005483.2 | NP_005474.2 | 2 - 3 | 208 | 83 | |
| XM_011527605.2 | XP_011525907.1 | 2 - 3 | 275 | 83 | ||
| XM_011527606.2 | XP_011525908.1 | 2 - 3 | 275 | 83 | ||
| XM_011527607.2 | XP_011525909.1 | 2 - 3 | 275 | 83 | ||
| GenBank mRNA | BC041606.1 | - | 2 - 3 | 133 | 83 | |
| BC052620.1 | - | 2 - 3 | 207 | 83 | ||
| BC067093.1 | - | 2 - 3 | 209 | 6339027 | 83 | |
| BQ889365.1 | - | 2 - 3 | 208 | 83 | ||
| U20979.1 | - | 2 - 3 | 94 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000088.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000589648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000589648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001409.hg.1 | ENST00000301280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001409.hg.1 | ENST00000344143.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001409.hg.1 | ENST00000535117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001409.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001409.hg.1 | TC19000088.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000589648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000589648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146972.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146972.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146972.hg.1 | TC1900006691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900146972.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146972.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146970.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146970.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146970.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146970.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146970.hg.1 | TC1900006691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900146970.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146970.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146970.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146970.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146971.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146971.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146971.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146971.hg.1 | TC1900006691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900146971.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146971.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146971.hg.1 | NM_005483.2 | Refseq |

CHAF1A
Genomic Map