Assay Details
| Gene Symbol: GIMAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018326.2 | NP_060796.1 | 1 - 2 | 69 | 151 | |
| GenBank mRNA | BX476689.1 | - | 1 - 2 | 70 | 193 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001004.hg
Additional Information:
This assay is in the 5'UTR region of the NM_018326.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07015266.hg.1 | ENST00000255945.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015266.hg.1 | ENST00000461940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015266.hg.1 | ENST00000466938.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015266.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015266.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015266.hg.1 | TC07001004.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015269.hg.1 | ENST00000255945.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015269.hg.1 | ENST00000461940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015269.hg.1 | ENST00000466938.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015269.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07015269.hg.1 | TC07001004.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700168734.hg.1 | TC0700009677.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000479232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009677.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | ENST00000255945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | ENST00000461940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | ENST00000478135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | ENST00000494750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | NM_018326.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700168730.hg.1 | TC0700009677.hg.1 | Clariom D TC |

GIMAP4
Genomic Map