Assay Details
| Gene Symbol: LRCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020871.3 | NP_065922.3 | 15 - 16 | 1774 | 128 | |
| XM_006724724.3 | XP_006724787.2 | 15 - 16 | 1756 | 128 | ||
| GenBank mRNA | AB040928.1 | - | 11 - 12 | 955 | 128 | |
| AK022128.1 | - | 7 - 8 | 530 | 128 | ||
| BC125223.1 | - | 3 - 4 | 181 | 128 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001291.hg.1
Set Membership: |
|

LRCH2
Genomic Map