Assay Details
| Gene Symbol: LRCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243963.1 | NP_001230892.1 | 16 - 17 | 1824 | 73 | |
| NM_020871.3 | NP_065922.3 | 17 - 18 | 1875 | 73 | ||
| XM_006724724.3 | XP_006724787.2 | 17 - 18 | 1857 | 73 | ||
| GenBank mRNA | AB040928.1 | - | 13 - 14 | 1056 | 73 | |
| AK022128.1 | - | 9 - 10 | 631 | 73 | ||
| BC125223.1 | - | 5 - 6 | 282 | 73 | ||
| BC125224.1 | - | 16 - 17 | 1817 | 40124466 | 73 | |
| BX647193.1 | - | 17 - 18 | 1875 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001291.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017464.hg.1 | ENST00000317135.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017464.hg.1 | ENST00000536655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017464.hg.1 | ENST00000538422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017464.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017464.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017464.hg.1 | TC0X001291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000607417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000607417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171938.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171938.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171938.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171938.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171938.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171938.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171938.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171938.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171938.hg.1 | TC0X00010559.hg.1 | Clariom D TC |

LRCH2
Genomic Map