Assay Details
| Gene Symbol: LRCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243963.1 | NP_001230892.1 | 7 - 8 | 1119 | 91 | |
| NM_020871.3 | NP_065922.3 | 7 - 8 | 1119 | 91 | ||
| XM_006724724.3 | XP_006724787.2 | 7 - 8 | 1122 | 91 | ||
| XM_017029696.1 | XP_016885185.1 | 7 - 8 | 1128 | 91 | ||
| XM_017029697.1 | XP_016885186.1 | 7 - 8 | 1129 | 91 | ||
| GenBank mRNA | AB040928.1 | - | 3 - 4 | 300 | 91 | |
| BC125224.1 | - | 7 - 8 | 1112 | 40124466 | 91 | |
| BX647193.1 | - | 7 - 8 | 1119 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001291.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017475.hg.1 | ENST00000317135.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017475.hg.1 | ENST00000538422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017475.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017475.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017475.hg.1 | TC0X001291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000607417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000607417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171953.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171953.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171953.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171953.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171953.hg.1 | ENST00000317135.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171953.hg.1 | ENST00000538422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171953.hg.1 | NM_001243963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171953.hg.1 | NM_020871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171953.hg.1 | TC0X00010559.hg.1 | Clariom D TC |

LRCH2
Genomic Map