| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000457332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000457332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000460032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000460032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000460032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000487075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000487075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000487075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
ENST00000487075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001513.hg.1 |
ENST00000253039.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001513.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001513.hg.1 |
TC0X000114.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000457332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000457332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000460032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000460032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000460032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000487075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000487075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000487075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
ENST00000487075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006803.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00146585.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146585.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146585.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00146585.hg.1 |
ENST00000253039.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146585.hg.1 |
ENST00000423068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146585.hg.1 |
NM_001415.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00146585.hg.1 |
TC0X00006803.hg.1 |
Clariom D TC |