Assay Details
| Gene Symbol: HOMER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277078.1 | NP_001264007.1 | 3 - 4 | 1361 | 126 | |
| NM_004272.4 | NP_004263.1 | 3 - 4 | 1361 | 126 | ||
| XM_017010059.1 | XP_016865548.1 | 3 - 4 | 2093 | 126 | ||
| GenBank mRNA | AF093262.1 | - | 2 - 3 | 299 | 126 | |
| AK223471.1 | - | 3 - 4 | 623 | 126 | ||
| AK312481.1 | - | 3 - 4 | 626 | 126 | ||
| AY189941.1 | - | 3 - 4 | 319 | 126 | ||
| BC015502.1 | - | 3 - 4 | 576 | 3904242 | 126 | |
| BC047438.1 | - | 3 - 4 | 1361 | 5294849 | 126 | |
| BT009846.1 | - | 2 - 3 | 299 | 126 | ||
| Y17829.1 | - | 3 - 4 | 374 | 126 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001522.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
phospholipase C-activating G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
positive regulation of signal transduction
behavioral response to cocaine
skeletal muscle fiber development
chemical homeostasis within a tissue
response to calcium ion
positive regulation of calcium ion transport
regulation of calcium ion import
regulation of store-operated calcium entry
regulation of cation channel activity
protein binding
G-protein coupled glutamate receptor binding
signaling adaptor activity
ion channel binding
skeletal muscle contraction
phospholipase C-activating G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
positive regulation of signal transduction
behavioral response to cocaine
skeletal muscle fiber development
chemical homeostasis within a tissue
response to calcium ion
positive regulation of calcium ion transport
regulation of calcium ion import
regulation of store-operated calcium entry
regulation of cation channel activity
protein binding
G-protein coupled glutamate receptor binding
signaling adaptor activity
ion channel binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05020871.hg.1 | ENST00000334082.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020871.hg.1 | ENST00000508576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020871.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020871.hg.1 | TC05001522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178028.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178028.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178028.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178028.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178028.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178028.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178028.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178028.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178028.hg.1 | TC0500011240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |

HOMER1
Genomic Map