| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027823.hg.1 |
ENST00000330909.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027823.hg.1 |
ENST00000402939.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027823.hg.1 |
ENST00000422949.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027823.hg.1 |
ENST00000483466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027823.hg.1 |
ENST00000497477.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027823.hg.1 |
NM_001033057.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027823.hg.1 |
NM_004742.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027823.hg.1 |
NM_015520.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027823.hg.1 |
TC03001528.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000330909.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000402939.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000460329.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000463103.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000472257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000483466.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000497477.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000611645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000621418.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
NM_001033057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
NM_004742.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
NM_015520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000330909.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000402939.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000460329.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000463103.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000472257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000483466.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000497477.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000611645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
ENST00000621418.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
NM_001033057.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
NM_004742.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
NM_015520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185535.hg.1 |
TC0300011403.hg.1 |
Clariom D TC |