Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927870 | LOC101927870 | -419.8kb | Anti-sense | Chr. 7 - 103445207 - 103514007 on NCBI Build GRCh38 | |
| Downstream | origin recognition complex subunit 5 | ORC5 |
ORC5L, ORC5P, ORC5T, PPP1R117 |
190.8kb | Sense | Chr. 7 - 104126341 - 104208047 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map