Assay Details
Gene Symbol: KMT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001324505.1 | NP_001311434.1 | 1 - 2 | 91 | 64 | |
XM_011538333.2 | XP_011536635.1 | 1 - 2 | 429 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001324505.1, XM_011538333.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
mitotic nuclear division
peptidyl-lysine monomethylation
histone H4-K20 methylation
regulation of DNA damage response, signal transduction by p53 class mediator
negative regulation of transcription, DNA-templated
cell division
regulation of signal transduction by p53 class mediator
p53 binding
transcription corepressor activity
protein binding
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H4-K20 specific)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
mitotic nuclear division
peptidyl-lysine monomethylation
histone H4-K20 methylation
regulation of DNA damage response, signal transduction by p53 class mediator
negative regulation of transcription, DNA-templated
cell division
regulation of signal transduction by p53 class mediator
p53 binding
transcription corepressor activity
protein binding
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H4-K20 specific)
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000462311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000462311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000462311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000478781.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000478781.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000478781.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000478781.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000537270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | ENST00000537270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000330479.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000402868.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000437519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000461103.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000462311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000462311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000462311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000478781.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000478781.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000478781.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000478781.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000485469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000537270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | ENST00000537270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009239.hg.1 | NM_020382.4 | Refseq |
