Assay Details
| Gene Symbol: GGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017023074.1 | XP_016878563.1 | 1 | 1622 | 154 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017023074.1 transcripts.
Set Membership: |
|

GGA2
Genomic Map