Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195328.1 | NP_001182257.1 | 1 - 2 | 171 | 109 | |
| GenBank mRNA | AK290505.1 | - | 1 - 2 | 150 | 109 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000060.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290505.1,NM_001195328.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
protein transport
regulation of protein localization
retrograde transport, endosome to Golgi
positive regulation of exocytosis
negative regulation by host of symbiont molecular function
GTPase activity
protein binding
GTP binding
GDP binding
small GTPase mediated signal transduction
protein transport
regulation of protein localization
retrograde transport, endosome to Golgi
positive regulation of exocytosis
negative regulation by host of symbiont molecular function
GTPase activity
protein binding
GTP binding
GDP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000809.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000809.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000809.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000809.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000809.hg.1 | TC0X000060.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006705.hg.1 | ENST00000415230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000805.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000805.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000805.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000805.hg.1 | TC0X000060.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000807.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000807.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000807.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000807.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000807.hg.1 | TC0X000060.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000810.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000810.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000810.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000810.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000810.hg.1 | TC0X000060.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006790.hg.1 | ENST00000449393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007272.hg.1 | ENST00000412737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000806.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000806.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000806.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000806.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000806.hg.1 | TC0X000060.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145532.hg.1 | TC0X00006656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0Y00006705.hg.1 | ENST00000415230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007272.hg.1 | ENST00000412737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145531.hg.1 | TC0X00006656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0Y00006790.hg.1 | ENST00000449393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145527.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145527.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145527.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145527.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145527.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145527.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145527.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145527.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145527.hg.1 | TC0X00006656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006656.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145528.hg.1 | TC0X00006656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | ENST00000243325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | ENST00000464506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | ENST00000618931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | NM_001195328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | NM_004251.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145529.hg.1 | TC0X00006656.hg.1 | Clariom D TC |

RAB9A
Genomic Map