| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000237.hg.1 |
ENST00000349721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000237.hg.1 |
ENST00000357248.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000237.hg.1 |
ENST00000382194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000237.hg.1 |
ENST00000382203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000237.hg.1 |
NM_003070.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000237.hg.1 |
NM_139045.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000237.hg.1 |
TC09000013.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
ENST00000349721.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
ENST00000357248.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
ENST00000382194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
ENST00000382203.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
NM_001289396.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
NM_001289397.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
NM_003070.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
NM_139045.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
ENST00000349721.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
ENST00000357248.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
ENST00000382194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
ENST00000382203.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
NM_001289396.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
NM_001289397.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
NM_003070.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
NM_139045.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144457.hg.1 |
TC0900006473.hg.1 |
Clariom D TC |