Assay Details
| Gene Symbol: MRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012219.4 | NP_036351.3 | 1 - 2 | 679 | 96 | |
| GenBank mRNA | AK124954.1 | - | 1 - 2 | 630 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000744.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK124954.1,NM_012219.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000289104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000423968.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000461114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000461114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000461114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000461114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000464896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000474559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000474559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000474559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000474559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000474559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000474559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000474559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000474559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000474559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000475711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000475711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000475711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000477784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000477784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000478647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000478647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000494949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000494949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000494949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000494949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000494949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000614350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | ENST00000621127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001085049.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_001252093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008945.hg.1 | NM_012219.4 | Refseq |

MRAS
Genomic Map