| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
ENST00000351718.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
ENST00000379022.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
ENST00000379024.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
ENST00000379028.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
ENST00000379032.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
ENST00000413765.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
ENST00000425651.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
ENST00000445979.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
ENST00000537765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
NM_001037132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
NM_001193582.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
NM_001193583.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
NM_001193584.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
NM_005010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027331.hg.1 |
TC07001751.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000351718.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000379022.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000379024.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000379028.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000413765.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000425651.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000613830.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_001037132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_001193582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_001193583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_001193584.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_005010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000351718.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000379022.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000379024.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000379028.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000413765.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000425651.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
ENST00000613830.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_001037132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_001193582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_001193583.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_001193584.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
NM_005010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700188285.hg.1 |
TC0700012230.hg.1 |
Clariom D TC |