Assay Details
| Gene Symbol: MYO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012223.3 | NP_036355.2 | 22 - 23 | 2633 | 73 | |
| GenBank mRNA | AJ001381.1 | - | 5 - 6 | 451 | 73 | |
| AJ001382.1 | - | 4 - 5 | 411 | 73 | ||
| AK000160.1 | - | 18 - 19 | 1967 | 73 | ||
More Information
Set Membership: |
|

MYO1B
Genomic Map