Assay Details
| Gene Symbol: UNC5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003728.3 | NP_003719.3 | 10 - 11 | 2091 | 90 | |
| XM_005263321.3 | XP_005263378.1 | 11 - 12 | 2148 | 90 | ||
| XM_017008789.1 | XP_016864278.1 | 11 - 12 | 8508 | 90 | ||
| XM_017008790.1 | XP_016864279.1 | 10 - 11 | 1736 | 90 | ||
| GenBank mRNA | AF055634.1 | - | 10 - 11 | 1892 | 90 | |
| AK290500.1 | - | 10 - 11 | 1902 | 90 | ||
| AK308421.1 | - | 11 - 12 | 1911 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001395.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04019421.hg.1 | ENST00000331502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019421.hg.1 | ENST00000453304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019421.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019421.hg.1 | TC04001395.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400177641.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400177641.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400177641.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400177641.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400177641.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400177641.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400177641.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400177641.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400177641.hg.1 | TC0400011359.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000453304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000504962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000506749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000513796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | ENST00000610318.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011359.hg.1 | NM_003728.3 | Refseq |

UNC5C
Genomic Map