Assay Details
| Gene Symbol: MSLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177355.2 | NP_001170826.1 | 2 - 3 | 99 | 109 | |
| GenBank mRNA | BC009272.2 | - | 2 - 3 | 98 | 3957372 | 109 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC009272.2, NM_001177355.2 transcripts.
Set Membership: |
|

MSLN
Genomic Map