Assay Details
| Gene Symbol: DHRS7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015510.4 | NP_056325.2 | 4 - 5 | 574 | 62 | |
| XM_005256595.4 | XP_005256652.2 | 5 - 6 | 1368 | 62 | ||
| XM_005256596.3 | XP_005256653.1 | 4 - 5 | 828 | 62 | ||
| XM_011523786.2 | XP_011522088.1 | 5 - 6 | 1368 | 62 | ||
| XM_017024425.1 | XP_016879914.1 | 4 - 5 | 686 | 62 | ||
| XM_017024426.1 | XP_016879915.1 | 5 - 6 | 913 | 62 | ||
| GenBank mRNA | AF151851.1 | - | 4 - 5 | 558 | 62 | |
| AL117567.1 | - | 4 - 5 | 561 | 62 | ||
| AY358498.2 | - | 3 - 4 | 522 | 62 | ||
| BC004126.2 | - | 4 - 5 | 548 | 3937286 | 62 | |
| BC009679.2 | - | 4 - 5 | 547 | 3878039 | 62 | |
| BI460496.1 | - | 4 - 5 | 575 | 62 | ||
| CR457124.1 | - | 4 - 5 | 530 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000295.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17003614.hg.1 | ENST00000346603.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003614.hg.1 | ENST00000395511.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003614.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003614.hg.1 | TC17000295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700151604.hg.1 | TC1700007254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700151606.hg.1 | TC1700007254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |

DHRS7B
Genomic Map