Assay Details
| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538838.1 | XP_011537140.1 | 4 - 5 | 629 | 64 | |
| XM_011538839.2 | XP_011537141.1 | 4 - 5 | 363 | 64 | ||
| XM_011538840.2 | XP_011537142.1 | 4 - 5 | 395 | 64 | ||
| XM_011538841.2 | XP_011537143.1 | 4 - 5 | 373 | 64 | ||
| XM_011538842.2 | XP_011537144.1 | 4 - 5 | 353 | 64 | ||
| XM_011538844.2 | XP_011537146.1 | 4 - 5 | 368 | 64 | ||
| XM_017020026.1 | XP_016875515.1 | 4 - 5 | 337 | 64 | ||
More Information
Set Membership: |
|

HVCN1
Genomic Map