Assay Details
| Gene Symbol: PECR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018441.5 | NP_060911.2 | 2 - 3 | 331 | 117 | |
| XM_011511478.2 | XP_011509780.1 | 1 - 2 | 132 | 117 | ||
| GenBank mRNA | AF212234.1 | - | 2 - 3 | 316 | 117 | |
| AF232009.1 | - | 2 - 3 | 321 | 117 | ||
| AJ250303.1 | - | 2 - 3 | 317 | 117 | ||
| AK315795.1 | - | 2 - 3 | 290 | 117 | ||
| BC002529.2 | - | 2 - 3 | 295 | 3139219 | 117 | |
| BU151224.1 | - | 2 - 3 | 331 | 117 | ||
| BX471043.1 | - | 2 - 3 | 325 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02005052.hg
Additional Information:
This assay is in the 5'UTR region of the XM_011511478.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044280.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044280.hg.1 | ENST00000497889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044280.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044280.hg.1 | TC02005052.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044281.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044281.hg.1 | ENST00000497889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044281.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044281.hg.1 | TC02005052.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228818.hg.1 | TC0200016768.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228819.hg.1 | TC0200016768.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |

PECR
Genomic Map