Assay Details
| Gene Symbol: DHRS7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015510.4 | NP_056325.2 | 1 - 2 | 92 | ||
| XM_017024426.1 | XP_016879915.1 | 2 - 3 | 92 | |||
| GenBank mRNA | AF151851.1 | - | 1 - 2 | 54 | 91 | |
| AL117567.1 | - | 1 - 2 | 92 | |||
| BI460496.1 | - | 1 - 2 | 92 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000295.hg
Additional Information:
This assay is in the 5'UTR region of the AF151851.1,AL117567.1,NM_015510.4,XM_017024426.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17003610.hg.1 | ENST00000346603.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003610.hg.1 | ENST00000395511.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003610.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003610.hg.1 | TC17000295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003606.hg.1 | ENST00000395511.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003606.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003606.hg.1 | TC17000295.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000581463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | ENST00000583388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007254.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | NM_015510.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | TC1700007254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000346603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000395511.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000577350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000578426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000579099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000579303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | ENST00000582161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700151598.hg.1 | NM_015510.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 186340 | - | - |

DHRS7B
Genomic Map