Assay Details
| Gene Symbol: SEC24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021982.2 | NP_068817.1 | 12 - 13 | 2076 | 156 | |
| XM_006714523.2 | XP_006714586.1 | 11 - 12 | 2277 | 156 | ||
| XM_017008961.1 | XP_016864450.1 | 12 - 13 | 1721 | 156 | ||
| XM_017008962.1 | XP_016864451.1 | 12 - 13 | 1493 | 156 | ||
| GenBank mRNA | AJ131244.1 | - | 12 - 13 | 1739 | 156 | |
| AK304060.1 | - | 12 - 13 | 2065 | 156 | ||
| AL137677.1 | - | 4 - 5 | 355 | 156 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000665.hg.1
Set Membership: |
|

SEC24A
Genomic Map