Assay Details
| Gene Symbol: SEC24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021982.2 | NP_068817.1 | 22 - 23 | 3463 | 109 | |
| XM_006714523.2 | XP_006714586.1 | 21 - 22 | 3664 | 109 | ||
| XM_017008961.1 | XP_016864450.1 | 22 - 23 | 3108 | 109 | ||
| XM_017008962.1 | XP_016864451.1 | 22 - 23 | 2880 | 109 | ||
| GenBank mRNA | AJ131244.1 | - | 22 - 23 | 3126 | 109 | |
| AK026545.1 | - | 9 - 10 | 1088 | 109 | ||
| AK304060.1 | - | 22 - 23 | 3452 | 109 | ||
| AL137677.1 | - | 14 - 15 | 1742 | 109 | ||
| BC009325.2 | - | 9 - 10 | 1105 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000665.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
antigen processing and presentation of peptide antigen via MHC class I
intracellular protein transport
ER to Golgi vesicle-mediated transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of low-density lipoprotein particle receptor biosynthetic process
COPII vesicle coating
positive regulation of protein secretion
positive regulation of cholesterol homeostasis
molecular_function
protein binding
zinc ion binding
antigen processing and presentation of peptide antigen via MHC class I
intracellular protein transport
ER to Golgi vesicle-mediated transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of low-density lipoprotein particle receptor biosynthetic process
COPII vesicle coating
positive regulation of protein secretion
positive regulation of cholesterol homeostasis
molecular_function
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05009217.hg.1 | ENST00000398844.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05009217.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05009217.hg.1 | TC05000665.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05009218.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05009218.hg.1 | TC05000665.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159754.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159754.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159754.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159754.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159754.hg.1 | TC0500008700.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500159755.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159755.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159755.hg.1 | TC0500008700.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500159753.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159753.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159753.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159753.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159753.hg.1 | TC0500008700.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 185535 | - | - |

SEC24A
Genomic Map