| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025577.hg.1 |
ENST00000281513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025577.hg.1 |
ENST00000441750.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025577.hg.1 |
NM_015909.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02025577.hg.1 |
TC02001592.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200185759.hg.1 |
ENST00000281513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200185759.hg.1 |
ENST00000442506.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200185759.hg.1 |
NM_015909.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200185759.hg.1 |
ENST00000281513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200185759.hg.1 |
ENST00000442506.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200185759.hg.1 |
NM_015909.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200185759.hg.1 |
TC0200011803.hg.1 |
Clariom D TC |